Phenol red
Glass bottle, 25 g
Catalog # TCIAP0100-25G
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:143-74-8
- Formula:C₁₉H₁₄O₅S
- MDL Number:MFCD00003552
- Molecular Weight:354.38 g/mol
- Merck Index:14,07247
- Melting Pt:> 300 °C
- Storage Temperature:Ambient
- EINECS:205-609-7
Documents
Specifications
Specification Test Results
- Appearance:Dark red to Brown powder to crystaline
- Absorbance:min. 0.530 (5mg/L, pH 8.4, 556.0 to 560.0 nm)
- Transition interval (pH):(yellow)6.8 to 8.4 (red)
Specifications
- Pack type:Glass bottle
- Pk:25 g



