Murexide
Glass bottle, 25 g
TCIAM0474-25G
: TCI
: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
- :3051-09-0
- :C₈H₈N₆O₆
- :MFCD00012777
- :284.19 g/mol
- :14,06306
- :300 °C
- :Ambient
- :221-266-6
- Appearance:Orange to Brown to Dark purple powder to crystal
- Lambda max.:520.0 to 524.0 nm (H2O)
- Absorbance (E1%1cm):min. 350 (H2O, 520.0 to 524.0 nm)
- Sensitiveness:to pass test
- Pack type:Glass bottle
- Pk:25 g



