Murexide
Glass bottle, 1 g
Catalog # TCIAM0474-1G
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
- CAS Number:3051-09-0
- Formula:C₈H₈N₆O₆
- MDL Number:MFCD00012777
- Molecular Weight:284.19 g/mol
- Merck Index:14,06306
- Melting Pt:300 °C
- Storage Temperature:Ambient
- EINECS:221-266-6
Documents
Specifications
Specification Test Results
- Appearance:Orange to Brown to Dark purple powder to crystal
- Lambda max.:520.0 to 524.0 nm (H2O)
- Absorbance (E1%1cm):min. 350 (H2O, 520.0 to 524.0 nm)
- Sensitiveness:to pass test
Specifications
- Pack type:Glass bottle
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



