Order Entry
Denmark
Orders LinkContactUsLinkComponent
Hydroxyurea ≥97.0% (by total nitrogen basis)

Hydroxyurea ≥97.0% (by total nitrogen basis)

Glass bottle, 5 g
Catalog # TCIAH0310-5G
Health
Supplier:   TCI
Hydroxyurea ≥97.0% (by total nitrogen basis)
Catalog # TCIAH0310-5G
Supplier:  TCI
CAS Number:  127-07-1
Hydroxyurea ≥97.0% (by total nitrogen basis)
Hydroxyurea ≥97.0% (by total nitrogen basis)

Glass bottle, 5 g

CAS Number:  127-07-1
Price
148,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  • CAS Number:127-07-1
  • Formula:CH₄N₂O₂
  • MDL Number:MFCD00007943
  • Molecular Weight:76.05 g/mol
  • Merck Index:14,04848
  • Boiling Pt:296 °C (1013 hPa)
  • Melting Pt:142…144 °C
  • Storage Temperature:Ambient
  • EINECS:204-821-7
  • UN:2811
  • ADR:6.1,III
Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 97.0 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g

Specifications