Order Entry
Denmark
ContactUsLinkComponent
NanoLC-Ultra 2Dplus system. Dual Gradient pump, 1-10 ?l/min for both gradients, and loading pump for 1-30 ?l/min. Max. pressure 10, 000 psi. Temperature controlled column compartment with two 10 port column switching valves. Includes NanoLC start-up 1 * 1 items
Catalog # 903-0609
Supplier:  EKSIGENT
NanoLC-Ultra 2Dplus system. Dual Gradient pump, 1-10 ?l/min for both gradients, and loading pump for 1-30 ?l/min. Max. pressure 10, 000 psi. Temperature controlled column compartment with two 10 port column switching valves. Includes NanoLC start-up 1 * 1 items
Catalog # 903-0609
Supplier:  EKSIGENT
Frequently Bought Together