Order Entry
Denmark
ContactUsLinkComponent
NanoLC-Ultra 2Dplus system. Dual Gradient pump, 50-500 nl/min for both gradients, and loading pump for 1-30 ?l/min. Max. pressure 1 0,000 psi. Temperature controlled column compartment with two 10 port column switching valves. Includes NanoLC start- 1 * 1 items
  903-0610
 :  EKSIGENT
NanoLC-Ultra 2Dplus system. Dual Gradient pump, 50-500 nl/min for both gradients, and loading pump for 1-30 ?l/min. Max. pressure 1 0,000 psi. Temperature controlled column compartment with two 10 port column switching valves. Includes NanoLC start- 1 * 1 items
  903-0610
 :  EKSIGENT
Frequently Bought Together