Order Entry
Denmark
ContactUsLinkComponent
NanoLC-Ultra 1Dplus system. Gradient pump for 50-500 nl/min and loading pump for 1-30 ?l/min. Max. pressure 10,000 psi. Temperature controlled column compartment with one 10 port column switching valve. Includes NanoLC start-up kit (p/n 801-00043) an 1 * 1 items
Catalog # 903-0618
Supplier:  EKSIGENT
NanoLC-Ultra 1Dplus system. Gradient pump for 50-500 nl/min and loading pump for 1-30 ?l/min. Max. pressure 10,000 psi. Temperature controlled column compartment with one 10 port column switching valve. Includes NanoLC start-up kit (p/n 801-00043) an 1 * 1 items
Catalog # 903-0618
Supplier:  EKSIGENT
Frequently Bought Together