Order Entry
Denmark
ContactUsLinkComponent
NanoLC-Ultra 2D system. Dual Gradient pump: 1-10 ?l/min for gradient 1, 50-500 nl/min for gradient 2. Max. pressure 10,000 psi. Tem perature controlled column compartment with one 10 port column switching valve. Includes NanoLC start-up kit (p/n 801- 1 * 1 items
  903-0616
 :  EKSIGENT
NanoLC-Ultra 2D system. Dual Gradient pump: 1-10 ?l/min for gradient 1, 50-500 nl/min for gradient 2. Max. pressure 10,000 psi. Tem perature controlled column compartment with one 10 port column switching valve. Includes NanoLC start-up kit (p/n 801- 1 * 1 items
  903-0616
 :  EKSIGENT
Frequently Bought Together