Order Entry
Denmark
Orders LinkContactUsLinkComponent
VWR® Buffer Solutions, Secondary Standards

VWR® Buffer Solutions, Secondary Standards

4,005, ±0,010, 25 °C, Plastic bottle, 500 mL
Catalog # 84582.260
Supplier:   VWR Chemicals
VWR® Buffer Solutions, Secondary Standards
Catalog # 84582.260
Supplier:  VWR Chemicals
CAS Number:  
VWR® Buffer Solutions, Secondary Standards
VWR® Buffer Solutions, Secondary Standards

4,005, ±0,010, 25 °C, Plastic bottle, 500 mL

CAS Number:  
Price
449,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents
  • Accurate to ±0,010 pH units (except buffer 12,454 with accuracy ±0,050)
  • Solution standardised at 25 °C
  • Fully traceable for optimum pH calibration (traceable to NIST, DIN 19266 and IUPAC)
  • Temperature dependence data is printed on the labels as are batch number and expiry date
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0,010
  • pH value:
    4,005

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.