Order Entry
Denmark
Orders LinkContactUsLinkComponent
Pendulum hardness tester
Pendulum hardness tester
Persoz pendulum, stainless steel, 500 g, SP0510
Catalog # SIMESP0510
Supplier:   Industrial Physics
Pendulum hardness tester
Catalog # SIMESP0510
Supplier:  Industrial Physics
Pendulum hardness tester
Pendulum hardness tester
Persoz pendulum, stainless steel, 500 g, SP0510
Price
3.740,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel.
  • Easy menu driven interface with jog dial
  • Design 'solid supports' to level the instrument
  • Integrated storage of glass plate and pendulum
  • Menu driven calibration menu eases both Persoz and König method
  • The end of a test is indicated by a visual and acoustic signal
The pendulum hardness test is based on the principle that the amplitude of oscillation of a pendulum touching a surface decreases more rapidly the softer the surface. It has a wide range of applications areas including laboratories, coating industry, paint industry, and the automotive industry.
Certifications: ISO 1522 (2005) standard for the correct execution of the test.
Documents
Specifications
  • Model:
    SP0510
  • Description:
    Persoz pendulum, stainless steel, 500 g
  • Seq:
    04