Order Entry
Denmark
Orders LinkContactUsLinkComponent
Coating thickness testers, PosiTector® 6000
Microprobe, N90S, 0 - 625 µm (±0,5 +1 % (0 - 100) ; ±2 +3 % (over 100)) for non conductive layers on NFe metals
  SIMELD6108
Coating thickness testers, PosiTector® 6000
  SIMELD6108
Coating thickness testers, PosiTector® 6000
Coating thickness testers, PosiTector® 6000
Microprobe, N90S, 0 - 625 µm (±0,5 +1 % (0 - 100) ; ±2 +3 % (over 100)) for non conductive layers on NFe metals
7.860,00 kr
1 * 1 Items
This coating thickness gauge is a fully electronic apparatus, that uses magnetic and eddy current principles, to measure coating thickness on both ferrous (Fe) and non ferrous (NFe) metals.
  • Fast measurement for quick inspection
  • Flip display enables right side up viewing
  • USB port for fast connection to a PC with continuous power
 : Software Ideal Finish Analysis is available on request. With two user levels Ideal Finish Analysis offers user friendly reporting functions for standard production work, as well as advanced calculations for in depth analysis. For the complete range of available probes, please contact your local Avantor agency. Please order probes separately.
 : Supplied with plastic shims, rubber holster with belt clip and wrist strap, three batteries, instructions, nylon carrying case with shoulder strap, certificate of calibration traceable to NIST and USB cable.
  • Model:
    N90S
  • Range:
    0 - 625 µm (±0,5 +1 % (0 - 100) ; ±2 +3 % (over 100)) for non conductive layers on NFe metals
  • Description:
    Microprobe
  • Seq:
    126
Accessories
  •  
    Description
  • SIMELD5505
    Infrared printer
    1 * 1 Items - 3.270,00 kr
  •  
    Description
  • SIMELD6015
    USB cable
    1 * 1 Items - 903,00 kr
  •  
    Description
  • SIMELD5508
    Bluetooth printer
    1 * 1 Items - 4.560,00 kr
  •  
    Description
    Range
  • 649-0022
    Coating thickness calibration standard
    0; 15; 40; 100 µm
    1 * 1 Items - 4.250,00 kr
  •  
    Description
    Range
  • 649-0023
    Coating thickness calibration standard
    0; 75; 250; 1500 µm
    1 * 1 Items - 4.250,00 kr
  •  
    Description
    Range
  • 649-0025
    Coating thickness calibration standard
    75; 125; 250; 500 µm
    1 * 1 Items - 4.250,00 kr
  •  
    Description
  • SIMELD6016
    PosiTector smartlink, Defelsko
    1 * 1 Items - 5.380,00 kr
  •  
    Description
  • SIMELD5407
    CAL P1 standard
    1 * 1 Items - 2.360,00 kr
  •  
    Description
  • SIMELD5408
    CAL P2 standard
    1 * 1 Items - 2.360,00 kr
  •  
    Description
  • SIMELD5409
    CAL P3 standard
    1 * 1 Items - 2.360,00 kr
  •  
    Description
  • SIMELD5410
    CAL P4 standard
    1 * 1 Items - 2.360,00 kr
  •  
    Description
    Model
    Range
  • SIMELD6017
    90° angle probe
    FRS
    0 - 1500 µm (±1 +1 % (0 - 50) ; ±2 +1 % (over 50)) for NFe metals
    1 * 1 Items - 5.380,00 kr
  •  
    Description
    Model
    Range
  • SIMELD6025
    Probe
    F
    0 - 1500 µm (±1 +1 % (0 - 50) ; ±2 +1 % (over 50)) for NFe metals
    1 * 1 Items - 3.830,00 kr
  •  
    Description
    Model
    Range
  • SIMELD6020
    Probe
    FKS
    0 - 13 000 mm (±0,02 +3 % (0 - 13)) for thick NFe metals
    1 * 1 Items - 6.530,00 kr
  •  
    Description
    Model
    Range
  • SIMELD6023
    Built-in detachable probe
    FNTS
    0 - 6000 mm (±0,01 +1 % (0 - 2,5) ; ±0,01 +3 % (over 2,5 mm)) for all metals
    1 * 1 Items - 9.780,00 kr
  •  
    Description
    Model
    Range
  • SIMELD6022
    90° angle probe
    FNRS
    0 - 1500 µm (±1 +1 % (0 - 50) ; ±2 +1 % (over 50)) for all metals
    1 * 1 Items - 9.210,00 kr
  •  
    Description
    Model
    Range
  • SIMELD6018
    Built-in detachable probe
    FTS
    0 - 6000 mm (±0,01 +1 % (0 - 2,5) ; ±0,01 +3 % (over 2,5)) for non-magnetic coatings over ferrous metals (steel and iron)
    1 * 1 Items - 6.010,00 kr
  •  
    Description
    Model
    Range
  • SIMELD6024
    Probe
    NKS
    0 - 13 000 mm (±0,02 +3 % (0 - 13)) for non conductive NFe metals
    1 * 1 Items - 6.730,00 kr