Order Entry
Denmark
ContactUsLinkComponent
15 COMPOSES DANS HNO3 5% Al 50mg/l As 50mg/l Ba 50mg/l Cd 50mg/l Co 50mg/l Cr 50mg/l Cu 50mg/l Mn 50mg/l Ni 50mg/l Pb 50mg/l Se 50mg/l Mo 50mg/l Sr 50mg/l Zn 50mg/l K 500mg/l 1 * 250 mL
Catalog # 89145.230
15 COMPOSES DANS HNO3 5% Al 50mg/l As 50mg/l Ba 50mg/l Cd 50mg/l Co 50mg/l Cr 50mg/l Cu 50mg/l Mn 50mg/l Ni 50mg/l Pb 50mg/l Se 50mg/l Mo 50mg/l Sr 50mg/l Zn 50mg/l K 500mg/l 1 * 250 mL
Catalog # 89145.230
Frequently Bought Together