Order Entry
Denmark
ContactUsLinkComponent
ekspert microLC 200 system consisting of a binary gradient pumping system configured for 5-50 uL/min flow rate, remotely mounted col umn oven, electrically-actuated low dispersion 6-port injection valve, and PAL DLW-HTC-xt autosampler with a six-plat 1 * 1 items
  903-0602
 :  EKSIGENT
undefined
ekspert microLC 200 system consisting of a binary gradient pumping system configured for 5-50 uL/min flow rate, remotely mounted col umn oven, electrically-actuated low dispersion 6-port injection valve, and PAL DLW-HTC-xt autosampler with a six-plat 1 * 1 items
  903-0602
 :  EKSIGENT
Frequently Bought Together