Order Entry
Denmark
Orders LinkContactUsLinkComponent
VWR® Redox Electrodes
RD 223 (redox), BNC, 1 m cable, Gel, Glass, 12×120 mm
  662-1409
VWR® Redox Electrodes
  662-1409
VWR® Redox Electrodes
VWR® Redox Electrodes
RD 223 (redox), BNC, 1 m cable, Gel, Glass, 12×120 mm
1.050,00 kr
1 * 1 Items
  • Connection:
    BNC, 1 m cable
  • Ø×L:
    12×120 mm
  • Description:
    RD 223 (redox)
  • Material:
    Glass
  • Electrolyte:
    Gel