Order Entry
Denmark
ContactUsLinkComponent
PRODUCT FOR RNA RELATE APPLICATION, DSRNA MARKER, THE DSRNA MARKER CONSISTS OF TEN DOUBLE-STRANDED RNAS, 10, 20, 30, 50, 100, 200, 3 00, 400, 500 AND 1,000 BASE PAIRS. THE DSRNA MARKER IS AN IDEAL SIZE MARKER FOR DETERMINATING SIZES OF DOUBLE-STRANDE 1 * 25 µG
Catalog # ABNOR0005
Supplier:  Abnova
PRODUCT FOR RNA RELATE APPLICATION, DSRNA MARKER, THE DSRNA MARKER CONSISTS OF TEN DOUBLE-STRANDED RNAS, 10, 20, 30, 50, 100, 200, 3 00, 400, 500 AND 1,000 BASE PAIRS. THE DSRNA MARKER IS AN IDEAL SIZE MARKER FOR DETERMINATING SIZES OF DOUBLE-STRANDE 1 * 25 µG
Catalog # ABNOR0005
Supplier:  Abnova
Frequently Bought Together