Safety helmets, Concept
Safety helmet, vented, without ratchet, EU, Black
Catalog # 111-0865
Supplier: CENTURION SAFETY PRODUCTS
Jump to:
Product Details & Documents
Lightweight, comfortable and modern protective helmet made from ABS (acrylonitrile butadiene styrene) with large forehead area featuring an imprint for the company logo.
- With extended nape area
- Six point terylene inner cradle
- Highly absorbent hydro-flock sweatband
- With increased side rigidity (additional lateral deformity test)
- Wide ventilation slots
- Without ratchet, with full peak
Size: 510 to 630 mm
Weight: 325 g
EN 397
: EN 397
Specifications
- Colour:Black
- Description:Safety helmet, vented, without ratchet, EU
- Seq:03



