Stearic acid
Glass bottle for solids, 10 g
CAYM10011298-10
: Octadecanoic acid, Octadecylic acid
- :57-11-4
- :C₁₈H₃₆O₂
- :MFCD00002752
- :284.48 g/mol
- :13,08882
- :361 °C (1013 hPa)
- :67 °C
- :0.94 g/cm³ (20 °C)
- :196 °C (closed cup)
- :Refrigerator
- :200-313-4
- Pack type:Glass bottle for solids
- Pk:10 g