Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories for DOSE IT
Accessories for DOSE IT
Pipette tubing connector for 2 - 6 mm ID silicone tubing, autoclavable, - mm
Catalog # INTG171077
Supplier:   INTEGRA Biosciences
Accessories for DOSE IT
Catalog # INTG171077
Accessories for DOSE IT
Accessories for DOSE IT
Pipette tubing connector for 2 - 6 mm ID silicone tubing, autoclavable, - mm
Price
496,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
Documents
Specifications
  • Description:
    Pipette tubing connector for 2 - 6 mm ID silicone tubing, autoclavable
  • Ø int.:
    - mm