Accessories for DOSE IT
Aspiration, dispensing tube, stainless steel, length: 10 cm, 2 mm
Catalog # INTG171052
Supplier: INTEGRA Biosciences
Jump to:
Product Details & Documents
Specifications
- Description:Aspiration, dispensing tube, stainless steel, length: 10 cm
- Ø int.:2 mm



