Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Accessories for DOSE IT

Silicone tubing set for pumphead, length: 3 m, 8 mm
Catalog # 710-1042
Supplier:   INTEGRA Biosciences
Accessories for DOSE IT
Catalog # 710-1042
Accessories for DOSE IT
Accessories for DOSE IT

Silicone tubing set for pumphead, length: 3 m, 8 mm

Price
1.170,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

 

Specifications
  • Description:
    Silicone tubing set for pumphead, length: 3 m
  • Ø int.:
    8 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.