Order Entry
Denmark
Orders LinkContactUsLinkComponent
DIPSO 99%

DIPSO 99%

Glass bottle for solids, 250 g
Catalog # H56187.30
DIPSO 99%
Catalog # H56187.30
CAS Number:  68399-80-4
DIPSO 99%
DIPSO 99%

Glass bottle for solids, 250 g

CAS Number:  68399-80-4
Price
6.540,00 kr
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 68399-80-4 Molecular Formula: C7H17NO6S Formula Weight: 261.29 Beilstein Reference: 2105454 MDL No.: MFCD00038353

Synonyms:  3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulphonic acid, DIPSO, 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulphonic acid
  • CAS Number:68399-80-4
  • Formula:C₇H₁₇NO₆S
  • MDL Number:MFCD00038353
  • Molecular Weight:261.29 g/mol
  • Storage Temperature:Ambient
  • EINECS:269-992-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.