Order Entry
Denmark
Orders LinkContactUsLinkComponent
DIPSO 99%

DIPSO 99%

Glass bottle for solids, 100 g
Catalog # H56187.22
DIPSO 99%
Catalog # H56187.22
CAS Number:  68399-80-4
DIPSO 99%
DIPSO 99%

Glass bottle for solids, 100 g

CAS Number:  68399-80-4
Price
3.120,00 kr
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 68399-80-4 Molecular Formula: C7H17NO6S Formula Weight: 261.29 Beilstein Reference: 2105454 MDL No.: MFCD00038353

Synonyms:  3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulphonic acid, DIPSO, 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulphonic acid
  • CAS Number:68399-80-4
  • Formula:C₇H₁₇NO₆S
  • MDL Number:MFCD00038353
  • Molecular Weight:261.29 g/mol
  • Storage Temperature:Ambient
  • EINECS:269-992-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications