Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Insulate containers, with locking clamps

15,5 l, 230 mm, 300 mm
  BOCH8087
 :   BOCHEM
Insulate containers, with locking clamps
  BOCH8087
 :  BOCHEM
Insulate containers, with locking clamps

15,5 l, 230 mm, 300 mm

 
2.480,00 kr
   
1 * 1 Items
 

Stainless steel 18/10.

  • With three locking clamps, lid and silicone gasket
  • Valve in the lid compensates possible over and under pressure
  • Very good chemical and temperature resistance
 
  • Capacity:
    15,5 l
  • Height:
    230 mm
  • Seq:
    03
  • Int.Ø:
    300 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.