Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

VWR®, Test Tubes Racks

White, 16 (4×4), 25 mm, 123,9×123,9×74 mm
  212-0395
VWR®, Test Tubes Racks
  212-0395
VWR®, Test Tubes Racks
VWR®, Test Tubes Racks

White, 16 (4×4), 25 mm, 123,9×123,9×74 mm

 
168,00 kr
   
1 * 1 Items
 

Nylon, glass fibre reinforced.

  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
 
  • For tube Ø:
    25 mm
  • No. of Holes:
    16 (4×4)
  • Colour:
    White
  • Seq:
    06
  • W×D×H:
    123,9×123,9×74 mm