Order Entry
Denmark
Orders LinkContactUsLinkComponent
VWR®, Test Tubes Racks

VWR®, Test Tubes Racks

White, 36 (6×6), 16 mm, 131×131×70 mm
Catalog # 212-0389
Supplier:   VWR Collection
VWR®, Test Tubes Racks
Catalog # 212-0389
Supplier:  VWR Collection
VWR®, Test Tubes Racks
VWR®, Test Tubes Racks

White, 36 (6×6), 16 mm, 131×131×70 mm

Price
192,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Nylon, glass fibre reinforced.

  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
Specifications
  • For tube Ø:
    16 mm
  • No. of Holes:
    36 (6×6)
  • Colour:
    White
  • Seq:
    29
  • W×D×H:
    131×131×70 mm