Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

VWR®, Test Tubes Racks

White, 72 (6×12), 16 mm, 127,3×250,6×73 mm
  212-0411
VWR®, Test Tubes Racks
  212-0411
VWR®, Test Tubes Racks
VWR®, Test Tubes Racks

White, 72 (6×12), 16 mm, 127,3×250,6×73 mm

 
188,00 kr
   
1 * 1 Items
 

Nylon, glass fibre reinforced.

  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
 
  • For tube Ø:
    16 mm
  • No. of Holes:
    72 (6×12)
  • Colour:
    White
  • W×D×H:
    127,3×250,6×73 mm