Order Entry
Denmark
Orders LinkContactUsLinkComponent
Powder funnels, PP

Powder funnels, PP

250 mm, 50 mm
Catalog # SAMP8023P-250
Supplier:   Sampling Systems
Powder funnels, PP
Catalog # SAMP8023P-250
Supplier:  Sampling Systems
Powder funnels, PP
Powder funnels, PP

250 mm, 50 mm

Price
386,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Reusable powder funnel, made of PP, crevice free, with wide diameter discharge spout to enable powder to be discharged without catching.

  • Good chemical resistance
  • Good temperature resistance
  • Ideal for loading powder into drums, containers or kegs
Specifications
  • Funnel O-Ø:
    250 mm
  • Stem O-Ø:
    50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.