Order Entry
Denmark
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

10 mm, 100 mm
Catalog # SAMP8022P-100
Supplier:   Sampling Systems
Funnels with short stem
Catalog # SAMP8022P-100
Supplier:  Sampling Systems
Funnels with short stem
Funnels with short stem

10 mm, 100 mm

Price
52,40 kr
Log in for availability
1 * 1 Items
Product Details & Documents

These liquid funnels have PP construction. Crevice free.

  • Good chemical resistance
  • Good temperature resistance
  • Easy to clean
Specifications
  • Funnel O-Ø:
    100 mm
  • Stem O-Ø:
    10 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.