Order Entry
Denmark
ContactUsLinkComponent
Peipro is a dna transfection reagent for transient gene expression (tge) for the production of recombinant proteins and antibodies i n hek-293 and cho cells grown in serum-free media. 1 * 1 l
Catalog # 115-01L
Peipro is a dna transfection reagent for transient gene expression (tge) for the production of recombinant proteins and antibodies i n hek-293 and cho cells grown in serum-free media. 1 * 1 l
Catalog # 115-01L
Frequently Bought Together