Order Entry
Denmark
Orders LinkContactUsLinkComponent
VWR® Drum Keys
VWR® Drum Keys
Drum key to open DIN 32/45/54/80 caps
Catalog # 84544.001
Supplier:   VWR Chemicals
VWR® Drum Keys
Catalog # 84544.001
Supplier:  VWR Chemicals
VWR® Drum Keys
VWR® Drum Keys
Drum key to open DIN 32/45/54/80 caps
Price
162,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
A selection of tools to assist in opening IBCs, 200 litre drums, plus bottles and other containers with DIN 32, 45, 54 and 80 caps.
Specifications
  • Description:
    Drum key to open DIN 32/45/54/80 caps