Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Closures for test tubes and flasks

Identi-plug foam plugs, Ø 35 - 45 mm
  JAECL800-D
Closures for test tubes and flasks
  JAECL800-D
Closures for test tubes and flasks
Closures for test tubes and flasks

Identi-plug foam plugs, Ø 35 - 45 mm

 
779,00 kr
   
1 * 100 Items
 

Identy-plug.

Plastic/foam plugs provide a snug fit, yet permit air flow; springs back to original shape after repeated use; autoclavable and chemically resistant.

Cotton cheesecloth.

100% pure cotton, very absorbent and low linting; 28×24 thread count.

Bleached cotton balls.

Available large or medium.

 
  • Description:
    Identi-plug foam plugs, Ø 35 - 45 mm
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.