Order Entry
Denmark
ContactUsLinkComponent
Gel Documentation System with 4 Mega Pixel camera, lens feedback lens, filterwheel with UV filter and 2 copies of GeneTools analysis software 1 * 1 items
Catalog # SYNOGBOX/CHEMI-XR5
Gel Documentation System with 4 Mega Pixel camera, lens feedback lens, filterwheel with UV filter and 2 copies of GeneTools analysis software 1 * 1 items
Catalog # SYNOGBOX/CHEMI-XR5
Frequently Bought Together