Single channel pipettes, mechanical, variable volume, PIPETMAN®
Product Details & Documents
PIPETMAN®, recognized as the pipetting standard for its reliability and proven durability, is a fully adjustable, air-displacement pipette that uses disposable tips. PIPETMAN combines legendary accuracy, precision, and durability with easy pipetting and handling.
- Legendary performance
- Easy pipetting
- Built-to-last and convenient
- Simple to maintain
PIPETMAN pipettes come in a large range of volumes with reduced pipetting forces for hours of comfortable pipetting. Eight single channel models cover a volume range from 0.2 µl to 10 ml and eight multichannel models cover a volume range from 1 µl to 300 µl.
Accurate and precise, PIPETMAN air-displacement pipettes are recognized as one of the world standard. They come in a large range of volumes, including single and multichannel models, allowing you to handle various sample types with confidence. PIPETMAN feels like PIPETMAN® Classic but with lower forces for enhanced pipetting habits with a standard pipette. Made of PVDF and stainless steel, PIPETMAN offers premium durability. Built-to-last, it is the economical solution for years of pipetting. Routine cleaning and maintenance are all that is required to keep PIPETMAN in top condition for years. The PIPETMAN design allows for easy access to user-serviceable parts, along with an autoclavable tip holder and ejector.
Ordering information: Models equipped with a tip ejector are only sold with a stainless steel tip ejector. Plastic tip ejectors are available in the accessories. Please note: PIPETMAN P5000 and P10 ml do not have tip ejectors.
Delivery information: The Microvolume starter kit contains the three pipettes P2G (0,2 - 2 µl), P10G (1 - 10 µl) and P100G (10 - 100 µl), 96 tips racks of DFL10ST and DF100ST DIAMOND tips, three SINGLE™ pipette holders, 10 COLORIS™ ID clips, Gilson guide to pipetting, and three plastic ejectors. The Pipetman® G starter kit contains the three pipettes P20G (2 - 20 µl), P200G (20 - 200 µl), P1000G (100 - 1000 µl), 96 tips racks of D200 and D1000 DIAMOND tips, three SINGLE™ pipette holders, 10 COLORIS™ ID clips, Gilson guide to pipetting and three plastic ejectors.
Caution: Please note: PIPETMAN P5000G and P10 mL G do not have a tip ejector.
Specifikationer
- Colour code:Light pink
- Volume:10 - 100 µl
- With:Stainless steel ejector
- Description:P100
- Seq:04
- Accuracy:±0,35 - ±0,80 µl
- Imprecision:≤0,10 - ≤0,15 µl
- Recommended tips:D200 / DF100 µl
Accessories
Produktinformation
- Katalog #DescriptionColourTilgængelighedPrice
- GILSF161406Pipette holder, SINGLE™, for one Gilson pipette, can be stucked to any smooth, dry surface or fastened with screwsWhiteLog ind for tilgængelighed1 * 1 Items - 141,74 kr
Produktinformation
- Katalog #DescriptionColourTilgængelighedPrice
- GILSF161405Angled pipette stand, TRIO™, for 3 Gilson pipettes (except Pipetman Concept®)TranslucentLog ind for tilgængelighed1 * 1 Items - 828,06 kr
Produktinformation
- Katalog #DescriptionColourTilgængelighedPrice
- 612-1816Rotating pipette stand, CARROUSEL™, for 7 Gilson pipettes (except Pipetman Concept®)WhiteLog ind for tilgængelighed1 * 1 Items - 939,96 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- GILSF144983Hand-free microtube opener, THE JIMMY™, opens both snap-cap and screw-cap tubes, set of threeLog ind for tilgængelighed1 * 1 Each - 228,28 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- GILSF161303Identification clips, COLORIS™, yellow, for personalising pipettesLog ind for tilgængelighed1 * 10 Items - 117,12 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- GILSF161304Identification clips, COLORIS™, green, for personalising pipettesLog ind for tilgængelighed1 * 10 Items - 117,12 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- GILSF161305Identification clips, COLORIS™, blue, for personalising pipettesLog ind for tilgængelighed1 * 10 Items - 117,12 kr
Produktinformation
- Katalog #DescriptionCapacityColourTilgængelighedPrice
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- GILSF161486Blue PIPETMAN® comfort handle with large, more comfortable finger hook, ergonomically designed, for use with PIPETMAN Classic, Neo®, and G pipettesLog ind for tilgængelighed1 * 3 Items - 382,70 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- GILSF161499Stickers for personalising the PIPETMAN® comfort handle, 12 stickers/sheet, 10 sheetsLog ind for tilgængelighed1 * 120 Items - 144,72 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- GILSF107027Plastic ejector for P2/P10 with adapterLog ind for tilgængelighed1 * 1 Items - 117,87 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- GILSF161301Identification clips, COLORIS™, mixed colours, for personalising pipettesLog ind for tilgængelighed1 * 10 Items - 117,12 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- GILSF161302Identification clips, COLORIS™, red, for personalising pipettesLog ind for tilgængelighed1 * 10 Items - 117,12 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- GILSF161306Identification clips, COLORIS™, white, for personalising pipettesLog ind for tilgængelighed1 * 10 Items - 117,12 kr
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
Produktinformation
- Katalog #DescriptionTilgængelighedPrice
- GILSF161488White PIPETMAN® comfort handle with large, more comfortable finger hook, ergonomically designed, for use with PIPETMAN Classic, Neo®, and G pipettesLog ind for tilgængelighed1 * 3 Items - 382,70 kr