Single channel pipettes, mechanical, variable volume, PIPETMAN®
Product Details & Documents
PIPETMAN®, recognized as the pipetting standard for its reliability and proven durability, is a fully adjustable, air-displacement pipette that uses disposable tips. PIPETMAN combines legendary accuracy, precision, and durability with easy pipetting and handling.
- Legendary performance
- Easy pipetting
- Built-to-last and convenient
- Simple to maintain
PIPETMAN pipettes come in a large range of volumes with reduced pipetting forces for hours of comfortable pipetting. Eight single channel models cover a volume range from 0.2 µl to 10 ml and eight multichannel models cover a volume range from 1 µl to 300 µl.
Accurate and precise, PIPETMAN air-displacement pipettes are recognized as one of the world standard. They come in a large range of volumes, including single and multichannel models, allowing you to handle various sample types with confidence. PIPETMAN feels like PIPETMAN® Classic but with lower forces for enhanced pipetting habits with a standard pipette. Made of PVDF and stainless steel, PIPETMAN offers premium durability. Built-to-last, it is the economical solution for years of pipetting. Routine cleaning and maintenance are all that is required to keep PIPETMAN in top condition for years. The PIPETMAN design allows for easy access to user-serviceable parts, along with an autoclavable tip holder and ejector.
: Models equipped with a tip ejector are only sold with a stainless steel tip ejector. Plastic tip ejectors are available in the accessories. Please note: PIPETMAN P5000 and P10 ml do not have tip ejectors.
: The Microvolume starter kit contains the three pipettes P2G (0,2 - 2 µl), P10G (1 - 10 µl) and P100G (10 - 100 µl), 96 tips racks of DFL10ST and DF100ST DIAMOND tips, three SINGLE™ pipette holders, 10 COLORIS™ ID clips, Gilson guide to pipetting, and three plastic ejectors. The Pipetman® G starter kit contains the three pipettes P20G (2 - 20 µl), P200G (20 - 200 µl), P1000G (100 - 1000 µl), 96 tips racks of D200 and D1000 DIAMOND tips, three SINGLE™ pipette holders, 10 COLORIS™ ID clips, Gilson guide to pipetting and three plastic ejectors.
: Please note: PIPETMAN P5000G and P10 mL G do not have a tip ejector.
Specifications
- Colour code:Orange
- Volume:0,2 - 2 µl
- With:Stainless steel ejector
- Description:P2
- Seq:01
- Accuracy:±0,024 - ±0,030 µl
- Imprecision:≤0,012 - ≤0,014 µl
- Recommended tips:D10 / DL10 / DF10 / DFL10 µl
Accessories
Product Information
- Catalog #ColourAvailabilityPrice
- Pipette holder, SINGLE™, for one Gilson pipette, can be stucked to any smooth, dry surface or fastened with screwsPipette holder, SINGLE™, for one Gilson pipette, can be stucked to any smooth, dry surface or fastened with screwsGILSF161406WhiteLog in for availability1 * 1 Items - 141,74 kr
Product Information
- Catalog #ColourAvailabilityPrice
- Angled pipette stand, TRIO™, for 3 Gilson pipettes (except Pipetman Concept®)Angled pipette stand, TRIO™, for 3 Gilson pipettes (except Pipetman Concept®)GILSF161405TranslucentLog in for availability1 * 1 Items - 828,06 kr
Product Information
- Catalog #ColourAvailabilityPrice
- Rotating pipette stand, CARROUSEL™, for 7 Gilson pipettes (except Pipetman Concept®)Rotating pipette stand, CARROUSEL™, for 7 Gilson pipettes (except Pipetman Concept®)612-1816WhiteLog in for availability1 * 1 Items - 939,96 kr
Product Information
- Catalog #AvailabilityPrice
- Hand-free microtube opener, THE JIMMY™, opens both snap-cap and screw-cap tubes, set of threeHand-free microtube opener, THE JIMMY™, opens both snap-cap and screw-cap tubes, set of threeGILSF144983Log in for availability1 * 1 Each - 228,28 kr
Product Information
- Catalog #AvailabilityPrice
- Identification clips, COLORIS™, yellow, for personalising pipettesIdentification clips, COLORIS™, yellow, for personalising pipettesGILSF161303Log in for availability1 * 10 Items - 117,12 kr
Product Information
- Catalog #AvailabilityPrice
- Identification clips, COLORIS™, green, for personalising pipettesIdentification clips, COLORIS™, green, for personalising pipettesGILSF161304Log in for availability1 * 10 Items - 117,12 kr
Product Information
- Catalog #AvailabilityPrice
- Identification clips, COLORIS™, blue, for personalising pipettesIdentification clips, COLORIS™, blue, for personalising pipettesGILSF161305Log in for availability1 * 10 Items - 117,12 kr
Product Information
- Catalog #CapacityColourAvailabilityPrice
- Reagent reservoir
Product Information
- Catalog #AvailabilityPrice
- Blue PIPETMAN® comfort handle with large, more comfortable finger hook, ergonomically designed, for use with PIPETMAN Classic, Neo®, and G pipettesBlue PIPETMAN® comfort handle with large, more comfortable finger hook, ergonomically designed, for use with PIPETMAN Classic, Neo®, and G pipettesGILSF161486Log in for availability1 * 3 Items - 382,70 kr
Product Information
- Catalog #AvailabilityPrice
- Stickers for personalising the PIPETMAN® comfort handle, 12 stickers/sheet, 10 sheetsStickers for personalising the PIPETMAN® comfort handle, 12 stickers/sheet, 10 sheetsGILSF161499Log in for availability1 * 120 Items - 144,72 kr
Product Information
- Catalog #AvailabilityPrice
- Plastic ejector for P2/P10 with adapter
Product Information
- Catalog #AvailabilityPrice
- Plastic ejector for P20
Product Information
- Catalog #AvailabilityPrice
- Plastic ejector for P200
Product Information
- Catalog #AvailabilityPrice
- Plastic ejector for P1000
Product Information
- Catalog #AvailabilityPrice
- Identification clips, COLORIS™, mixed colours, for personalising pipettesIdentification clips, COLORIS™, mixed colours, for personalising pipettesGILSF161301Log in for availability1 * 10 Items - 117,12 kr
Product Information
- Catalog #AvailabilityPrice
- Identification clips, COLORIS™, red, for personalising pipettesIdentification clips, COLORIS™, red, for personalising pipettesGILSF161302Log in for availability1 * 10 Items - 117,12 kr
Product Information
- Catalog #AvailabilityPrice
- Identification clips, COLORIS™, white, for personalising pipettesIdentification clips, COLORIS™, white, for personalising pipettesGILSF161306Log in for availability1 * 10 Items - 117,12 kr
Product Information
- Catalog #AvailabilityPrice
- Plastic ejector for P100
Product Information
- Catalog #AvailabilityPrice
- White PIPETMAN® comfort handle with large, more comfortable finger hook, ergonomically designed, for use with PIPETMAN Classic, Neo®, and G pipettesWhite PIPETMAN® comfort handle with large, more comfortable finger hook, ergonomically designed, for use with PIPETMAN Classic, Neo®, and G pipettesGILSF161488Log in for availability1 * 3 Items - 382,70 kr