Order Entry
Denmark
Orders LinkContactUsLinkComponent
Blue Dextran loading dye
10 mL
  786-574
Blue Dextran loading dye
  786-574
 :  87915-38-6
Blue Dextran loading dye
Blue Dextran loading dye
10 mL
 :  87915-38-6
1.270,00 kr
1 * 10 mL
A proprietary mixture of Formaldehyde, EDTA and Blue Dextran that is designed for use in loading DNA samples onto denaturing polyacrylamide gels.
Commonly used in loading DNA samples onto denaturing polyacrylamide gels.
 :  Dextran, blue
  •  :87915-38-6
  •  :(C₂₉H₂₁N₇O₁₂S₃·3Na)n
  •  :MFCD00130661
  • Pk:
    10 mL