Microtube 2ml polypropylene skirted with screw cap assembled sterile 1 * 2.500 items
Catalog # GREI722361_2500
Supplier: Greiner Bio-One
Microtube 2ml polypropylene skirted with screw cap assembled sterile 1 * 2.500 items
Catalog # GREI722361_2500
Supplier: Greiner Bio-One
Frequently Bought Together
Serological pipettes, Stripette™
Supplier: Corning
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
VWR®, Cryo Boxes
Supplier: VWR Collection
Cryogenic tubes, CryoTubes™, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Cell Strainers
Supplier: VWR Collection
VWR®, Sealing films, Non Tacky, for qPCR, Storage and Crystallisation
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes and Screw Caps
Supplier: VWR Collection
Microplates, 96- and 384-well, Nunc™
Supplier: Thermo Fisher Scientific
Serological pipettes
Supplier: Greiner Bio-One
Multiple well cell culture plates, Costar®
Supplier: Corning
VWR® Cellulose Stoppers
Supplier: VWR Collection
Cell culture dishes, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Transfer pipettes, Samco™ Extra Long
Supplier: SAMCO
Accessories for MagBinder® Fit24 Nucleic Acid Purification System
Supplier: OMEGA BIO-TEK
Modules, Immuno™ LockWell™ Fluoronunc™/Luminunc™
Supplier: Thermo Fisher Scientific
Water sampling bottles, rectangular
Supplier: DELTALAB