Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Solid phase extraction cartridges, HyperSep™ C18

Cartridge, C18, 40 - 60 µm, 10 ml, 500 mg
  HYPE60108-786
Solid phase extraction cartridges, HyperSep™ C18
  HYPE60108-786
Solid phase extraction cartridges, HyperSep™ C18
Solid phase extraction cartridges, HyperSep™ C18

Cartridge, C18, 40 - 60 µm, 10 ml, 500 mg

 
2.290,00 kr
   
1 * 50 Items
 

These columns feature a highly retentive alkyl-bonded phase for non polar to moderately polar compounds.

  • Hydrophobic reversed phase
  • Retains most organic analytes from aqueous matrices
 
  • Column Format:
    Cartridge
  • Particle Size:
    40 - 60 µm
  • Capacity:
    10 ml
  • Seq:
    08
  • Bed weight:
    500 mg
  • Phase:
    C18