Order Entry
Denmark
Orders LinkContactUsLinkComponent
Dry buffer packs, JAW™

Dry buffer packs, JAW™

Tris buffered saline (TBS), JAW™, 20X, dry buffer pack for 20 l of 1X/ pack, 0,5 M Tris, 2,8 M sodium chloride and 60 mM potassium chloride, 7,25 - 7,55, 1
Catalog # RC-148
Supplier:   G-Biosciences
Dry buffer packs, JAW™
Catalog # RC-148
Supplier:  G-Biosciences
CAS Number:  
Dry buffer packs, JAW™

Tris buffered saline (TBS), JAW™, 20X, dry buffer pack for 20 l of 1X/ pack, 0,5 M Tris, 2,8 M sodium chloride and 60 mM potassium chloride, 7,25 - 7,55, 1

CAS Number:  
Price
833,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Premixed buffer powders that simply require adding water to generate a ready-to-use buffer.

Documents

Specifications

Specifications

  • Pk:
    1
  • Description:
    Tris buffered saline (TBS), JAW™, 20X, dry buffer pack for 20 l of 1X/ pack
  • pH value:
    7,25 - 7,55
  • Contents:
    0,5 M Tris, 2,8 M sodium chloride and 60 mM potassium chloride

Specifications