Dry buffer packs, JAW™
Phosphate buffered saline (PBS), JAW™, 10X, dry buffer pack for 10 l of 1X/ pack, 43 mM NaH₂PO₄, 1,37 M NaCl, 27mM KCl and 14 mM KH₂PO₄, 7,4, 2
Catalog # RC-147
Supplier: G-Biosciences
Jump to:
Product Details & Documents
Premixed buffer powders that simply require adding water to generate a ready-to-use buffer.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:2
- Description:Phosphate buffered saline (PBS), JAW™, 10X, dry buffer pack for 10 l of 1X/ pack
- pH value:7,4
- Contents:43 mM NaH₂PO₄, 1,37 M NaCl, 27mM KCl and 14 mM KH₂PO₄