Multi purpose bottles
Labels, self-adhesive, for multi purpose bottles
Catalog # RATI5922253
Supplier: RATIOLAB
Gå til:
Product Details & Documents
Polyethylene, blue translucent.
- Incl. lid with nonwoven fabric liner
- With graduations
- Light protection
- Wide neck diameter of 80 mm
Specifikationer
- Description:Labels, self-adhesive, for multi purpose bottles
- Seq:03