Order Entry
Denmark
Orders LinkContactUsLinkComponent
Multi purpose bottles

Multi purpose bottles

Lids for multipurpose collecting bottles, 2000 or 2500 ml capacity
Catalog # RATI5922204
Supplier:   RATIOLAB
Multi purpose bottles
Catalog # RATI5922204
Supplier:  RATIOLAB
Multi purpose bottles
Multi purpose bottles

Lids for multipurpose collecting bottles, 2000 or 2500 ml capacity

Price
Inquire For Price
Log in for availability
1 * 100 Items
Product Details & Documents

Polyethylene, blue translucent.

  • Incl. lid with nonwoven fabric liner
  • With graduations
  • Light protection
  • Wide neck diameter of 80 mm
Specifications
  • Description:
    Lids for multipurpose collecting bottles, 2000 or 2500 ml capacity
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.