Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Screw caps for jars

Screw cap, PP, 90 mm, White, Jar, 500 ml
  LAMA2458-2
 :   LAMAPLAST
Screw caps for jars
  LAMA2458-2
 :  LAMAPLAST
Screw caps for jars
Screw caps for jars

Screw cap, PP, 90 mm, White, Jar, 500 ml

 
15,20 kr
   
1 * 1 Items
 
  • For:
    Jar, 500 ml
  • Colour:
    White
  • Description:
    Screw cap, PP, 90 mm
  • Seq:
    04