Order Entry
Denmark
Orders LinkContactUsLinkComponent
Chloroform solution, NMR reference standard, Supelco®

Chloroform solution, NMR reference standard, Supelco®

Chloroform solution, 1% in acetone-d6 (99.9 atom % D), NMR reference standard, NMR tube size 3 mm x 8 in., 1
Catalog # 611778-1EA
Supplier:   Merck
Chloroform solution, NMR reference standard, Supelco®
Catalog # 611778-1EA
Supplier:  Merck
CAS Number:  
Chloroform solution, NMR reference standard, Supelco®
Chloroform solution, NMR reference standard, Supelco®

Chloroform solution, 1% in acetone-d6 (99.9 atom % D), NMR reference standard, NMR tube size 3 mm x 8 in., 1

CAS Number:  
Price
1.610,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
Specifications

Specifications

  • Concentration:
    1% in acetone-d6 (99.9 atom % D)
  • Pk:
    1
  • Standards:
    NMR reference standard
  • Description:
    Chloroform solution
  • Pk Info:
    NMR tube size 3 mm x 8 in.
  • Cat. No.:
    611778-1EA
  • Description:
    Chloroform solution

Specifications