Chloroform solution, NMR reference standard, Supelco®
Chloroform solution, 5% in acetone-d6 (99.9 atom % D), NMR reference standard, NMR tube size 5 mm x 8 in., 1
Catalog # 487759-1EA
Supplier: Merck
Chloroform solution, NMR reference standard, Supelco®
Catalog # 487759-1EA
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Concentration:5% in acetone-d6 (99.9 atom % D)
- Pk:1
- Standards:NMR reference standard
- Description:Chloroform solution
- Pk Info:NMR tube size 5 mm x 8 in.
- Cat. No.:487759-1EA
- Description:Chloroform solution



