Order Entry
Denmark
Orders LinkContactUsLinkComponent
Control units
Control units
MIXcontrol 20, with RS232 interface, CH-plug
Catalog # TMAG90210_CH
Control units
Catalog # TMAG90210_CH
Supplier:  
Control units
Control units
MIXcontrol 20, with RS232 interface, CH-plug
Discontinued
Product Details & Documents
Specifications
  • Description:
    MIXcontrol 20, with RS232 interface, CH-plug
  • Cat. No.:
    TMAG90210_CH
  • Model:
    MIXcontrol 20
  • Speed range:
    100 - 1600 min⁻¹
  • W×D×H:
    200×155×38 mm
  • Power input:
    20 W
  • Weight:
    1,9 kg