Order Entry
Denmark
ContactUsLinkComponent
COMPOSE A 10 MULTIELEMENTS DANS HCL 3%,HNO3 1% Al 1mg/l B 5mg/l Co 1mg/l Cr 0.5mg/l Cu 0.5mg/l Fe 1mg/l Mg 5mg/l Ni 0.5mg/l P 5mg/l Zn 0.5mg/l 1 * 250 mL
Catalog # CMPL0FB8.1.3C1N.L2
COMPOSE A 10 MULTIELEMENTS DANS HCL 3%,HNO3 1% Al 1mg/l B 5mg/l Co 1mg/l Cr 0.5mg/l Cu 0.5mg/l Fe 1mg/l Mg 5mg/l Ni 0.5mg/l P 5mg/l Zn 0.5mg/l 1 * 250 mL
Catalog # CMPL0FB8.1.3C1N.L2
Frequently Bought Together