Order Entry
Denmark
Orders LinkContactUsLinkComponent
Detergent removal system, DetergentOUT™ Tween®
Detergent removal system, DetergentOUT™ Tween®
DetergentOUT™ Tween®, Medi, 10
Catalog # 786-215
Supplier:   G-Biosciences
Detergent removal system, DetergentOUT™ Tween®
Catalog # 786-215
Supplier:  G-Biosciences
CAS Number:  
Detergent removal system, DetergentOUT™ Tween®
Detergent removal system, DetergentOUT™ Tween®
DetergentOUT™ Tween®, Medi, 10
CAS Number:  
Price
3.670,00 kr
Log in for availability
1 * 10 Items
Product Details & Documents
DetergentOUT™ Tween® is a spin column format detergent removal resin for polysorbate or Tween® detergents or surfactants.
DetergentOUT™ Tween® specifically removes polysorbate detergents without significant loss of proteins, dilution of the protein solution, or change to the buffer composition of the protein solution.
Specifications
Specifications
  • Pk:
    10
  • Description:
    DetergentOUT™ Tween®, Medi