Order Entry
Denmark
ContactUsLinkComponent
Air Jacketed CO2 Incubator with InCuSafe Stainless Steel-Copper Alloy 170 litre internal capacity (6.0 cu.ft) with 4 height adjusta ble shelves. External Dimensions: 620 x 710 x 900mm (wdh). Internal Dimensions: 490 x 523 x 665mm (wdh). Adjustable i 1 * 1 items
Catalog # 390-0588
Supplier:  PHC Corporation
undefined
Air Jacketed CO2 Incubator with InCuSafe Stainless Steel-Copper Alloy 170 litre internal capacity (6.0 cu.ft) with 4 height adjusta ble shelves. External Dimensions: 620 x 710 x 900mm (wdh). Internal Dimensions: 490 x 523 x 665mm (wdh). Adjustable i 1 * 1 items
Catalog # 390-0588
Supplier:  PHC Corporation
Frequently Bought Together