Order Entry
Denmark
Orders LinkContactUsLinkComponent
Graduated pipettes, type 1, Class AS, MBL

Graduated pipettes, type 1, Class AS, MBL

25 ml, 0,20 ml, 0,10 ± ml, Green
Katalog # 612-3944
Leverandør:   SciLabware
Graduated pipettes, type 1, Class AS, MBL
Katalog # 612-3944
Leverandør:  SciLabware
Graduated pipettes, type 1, Class AS, MBL
Graduated pipettes, type 1, Class AS, MBL

25 ml, 0,20 ml, 0,10 ± ml, Green

Price
181,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents

Soda lime glass.

  • Calibrated to deliver from zero at the top to any graduation line down to the shoulder
  • Graduations in permanent amber stain

DIN EN ISO 835

Certifications: DIN EN ISO 835.

Specifikationer
  • Colour code:
    Green
  • Capacity:
    25 ml
  • Division:
    0,20 ml
  • Tolerance:
    0,10 ± ml
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.