Order Entry
Denmark
Orders LinkContactUsLinkComponent
Maintenance solutions for electrodes

Maintenance solutions for electrodes

Pepsin-HCl for cleaning junctions with protein contamination, 250 ml, 250 mL
Catalog # METT51350100
Supplier:   Mettler - Toledo
Maintenance solutions for electrodes
Catalog # METT51350100
Supplier:  Mettler - Toledo
CAS Number:  
Maintenance solutions for electrodes

Pepsin-HCl for cleaning junctions with protein contamination, 250 ml, 250 mL

CAS Number:  
Price
275,00 kr
Log in for availability
1 * 250 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Pepsin-HCl for cleaning junctions with protein contamination, 250 ml

Specifications